2,4,5-Trimethyl-1H-pyrrole-3-carbaldehyde 95%
Lieferant: Apollo Scientific
OR918414-1G
OR918414-250MG
APOSOR918414-1GEA
464
EUR
APOSOR918414-1G
APOSOR918414-250MG
2,4,5-Trimethyl-1H-pyrrole-3-carbaldehyde 95%
2,4,5-Trimethyl-1H-pyrrole-3-carbaldehyde
Formel:
C₈H₁₁NO Molecular Weight: 137.179 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12827508 CAS-Nummer: 288089-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...