2-Cyano-4-methyl biphenyl 95%
Lieferant: Apollo Scientific
Synonyme:
4-Methyl-[1,1'-biphenyl]-2-carbonitrile
OR918479-10G
OR918479-1G
OR918479-5G
APOSOR918479-10GEA
132
EUR
APOSOR918479-10G
APOSOR918479-1G
APOSOR918479-5G
2-Cyano-4-methyl biphenyl 95%
2-Cyano-4-methyl biphenyl
Formel:
C₁₄H₁₁N Molecular Weight: 193.244 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06411093 CAS-Nummer: 64113-85-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...