5-Methoxy-2-methylphenol 95%

Lieferant: Apollo Scientific

OR918494-1G OR918494-5G
APOSOR918494-1GEA 79.9 EUR
APOSOR918494-1G APOSOR918494-5G
5-Methoxy-2-methylphenol 95%
5-Methoxy-2-methylphenol
Formel: C₈H₁₀O₂
Molecular Weight: 138,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12405727
CAS-Nummer: 20734-74-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR