2-Chloro-4-nitropyridin-3-ol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR918527-1G
OR918527-250MG
OR918527-5G
APOSOR918527-1GEA
114
EUR
APOSOR918527-1G
APOSOR918527-250MG
APOSOR918527-5G
2-Chloro-4-nitropyridin-3-ol 97%
2-Chloro-4-nitropyridin-3-ol
Formel:
C₅H₃ClN₂O₃ Molecular Weight: 174,542 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11847780 CAS-Nummer: 15128-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...