Thieno[2,3-c]pyridine-2-carboxylic acid 96%
Lieferant: Apollo Scientific
OR918586-1G
OR918586-250MG
APOSOR918586-1GEA
763
EUR
APOSOR918586-1G
APOSOR918586-250MG
Thieno[2,3-c]pyridine-2-carboxylic acid 96%
Thieno[2,3-c]pyridine-2-carboxylic acid
Formel:
C₈H₅NO₂S Molecular Weight: 179,196 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13659442 CAS-Nummer: 478149-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...