2-Chloro-6-hydroxy-4-methoxybenzaldehyde 95%

Lieferant: Apollo Scientific

OR918606-1G OR918606-250MG
APOSOR918606-1GEA 877 EUR
APOSOR918606-1G APOSOR918606-250MG
2-Chloro-6-hydroxy-4-methoxybenzaldehyde 95%
2-Chloro-6-hydroxy-4-methoxybenzaldehyde
Formel: C₈H₇ClO₃
Molecular Weight: 186,592 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23705594
CAS-Nummer: 116475-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR