5-Methoxy-1H-benzo[d]imidazole-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR918639-1G OR918639-250MG
APOSOR918639-1GEA 328 EUR
APOSOR918639-1G APOSOR918639-250MG
5-Methoxy-1H-benzo[d]imidazole-2-carboxylic acid 95%
5-Methoxy-1H-benzo[d]imidazole-2-carboxylic acid
Formel: C₉H₈N₂O₃
Molecular Weight: 192.171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09261054
CAS-Nummer: 887572-60-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR