4-(1-Hydroxy-ethyl)-benzoic acid 95%

Lieferant: Apollo Scientific

Synonyme: 4-(1-Hydroxyethyl)benzoic acid

OR918652-1G OR918652-250MG
APOSOR918652-1GEA 151 EUR
APOSOR918652-1G APOSOR918652-250MG
4-(1-Hydroxy-ethyl)-benzoic acid 95%
4-(1-Hydroxy-ethyl)-benzoic acid
Formel: C₉H₁₀O₃
Molecular Weight: 166.174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00870634
CAS-Nummer: 97364-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR