5-Iodoisoquinoline ≥98%

Lieferant: Apollo Scientific

Synonyme: DSRM-3716

OR918696-5G
APOSOR918696-5GEA 246 EUR
APOSOR918696-5G
5-Iodoisoquinoline ≥98%
5-Iodoisoquinoline
Formel: C₉H₆IN
Molecular Weight: 255,055 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226893
CAS-Nummer: 58142-99-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR