N-(3-Formyl-2,5-dimethyl-1H-pyrrol-1-yl)acetamide 95%
Lieferant: Apollo Scientific
OR918721-1G
OR918721-250MG
APOSOR918721-1GEA
400
EUR
APOSOR918721-1G
APOSOR918721-250MG
N-(3-Formyl-2,5-dimethyl-1H-pyrrol-1-yl)acetamide 95%
N-(3-Formyl-2,5-dimethyl-1H-pyrrol-1-yl)acetamide
Formel:
C₉H₁₂N₂O₂ Molecular Weight: 180,204 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09675014 CAS-Nummer: 932186-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...