6-(Ethoxycarbonyl)pyridine-2-carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 6-(Ethoxycarbonyl)picolinic acid

OR918772-10G OR918772-1G OR918772-5G
APOSOR918772-10GEA 235 EUR
APOSOR918772-10G APOSOR918772-1G APOSOR918772-5G
6-(Ethoxycarbonyl)pyridine-2-carboxylic acid 95%
6-(Ethoxycarbonyl)pyridine-2-carboxylic acid
Formel: C₉H₉NO₄
Molecular Weight: 195.172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00806154
CAS-Nummer: 21855-16-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR