1,2-Dimethoxy-4-ethylbenzene 95%

Lieferant: Apollo Scientific

Synonyme: 4-Ethyl-1,2-dimethoxybenzene

OR918848-1G OR918848-250MG
APOSOR918848-1GEA 683 EUR
APOSOR918848-1G APOSOR918848-250MG
1,2-Dimethoxy-4-ethylbenzene 95%
1,2-Dimethoxy-4-ethylbenzene
Formel: C₁₀H₁₄O₂
Molecular Weight: 166.217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00673001
CAS-Nummer: 5888-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR