4,4'-Dihydroxy-2,2'-bipyridine 95%
Lieferant: Apollo Scientific
Synonyme:
[2,2'-Bipyridine]-4,4'-diol
OR918861-1G
OR918861-250MG
APOSOR918861-1GEA
230
EUR
APOSOR918861-1G
APOSOR918861-250MG
4,4'-Dihydroxy-2,2'-bipyridine 95%
4,4'-Dihydroxy-2,2'-bipyridine
Formel:
C₁₀H₈N₂O₂ Molecular Weight: 188,183 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08704189 CAS-Nummer: 90770-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...