6-Bromo-1-methyl-1H-benzo[d][1,2,3]triazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR918875-1G
OR918875-250MG
APOSOR918875-1GEA
87
EUR
APOSOR918875-1G
APOSOR918875-250MG
6-Bromo-1-methyl-1H-benzo[d][1,2,3]triazole 97%
6-Bromo-1-methyl-1H-benzo[d][1,2,3]triazole
Formel:
C₇H₆BrN₃ Molecular Weight: 212,047 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11559050 CAS-Nummer: 1083181-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...