2-Ethoxyethanesulphonyl chloride 96%
Lieferant: Apollo Scientific
OR918902-1G
OR918902-5G
APOSOR918902-1GEA
139
EUR
APOSOR918902-1G
APOSOR918902-5G
2-Ethoxyethanesulphonyl chloride 96%
2-Ethoxyethanesulphonyl chloride
Formel:
C₄H₉ClO₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD10032392 CAS-Nummer: 69371-75-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...