Methyl isoquinoline-6-carboxylate 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 6-Isoquinolinecarboxylate
OR918920-1G
OR918920-250MG
APOSOR918920-1GEA
172
EUR
APOSOR918920-1G
APOSOR918920-250MG
Methyl isoquinoline-6-carboxylate 98%
Methyl isoquinoline-6-carboxylate
Formel:
C₁₁H₉NO₂ Molecular Weight: 187,195 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226961 CAS-Nummer: 173089-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...