1-(3-Methoxypyridin-2-yl)ethanone 95%
Lieferant: Apollo Scientific
OR918940-1G
OR918940-5G
APOSOR918940-1GEA
106
EUR
APOSOR918940-1G
APOSOR918940-5G
1-(3-Methoxypyridin-2-yl)ethanone 95%
1-(3-Methoxypyridin-2-yl)ethanone
Formel:
C₈H₉NO₂ Molecular Weight: 151.163 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18256469 CAS-Nummer: 379227-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...