3-Chloro-1H-pyrazolo[3,4-b]pyridine 95%

Lieferant: Apollo Scientific

OR918964-1G
APOSOR918964-1GEA 179 EUR
APOSOR918964-1G
3-Chloro-1H-pyrazolo[3,4-b]pyridine 95%
3-Chloro-1H-pyrazolo[3,4-b]pyridine
Formel: C₆H₄ClN₃
Molecular Weight: 153,569 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06254093
CAS-Nummer: 117007-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR