2-(Piperidin-1-yl)pyrimidine-5-carbaldehyde 95%

Lieferant: Apollo Scientific

OR918977-1G OR918977-250MG
APOSOR918977-1GEA 253 EUR
APOSOR918977-1G APOSOR918977-250MG
2-(Piperidin-1-yl)pyrimidine-5-carbaldehyde 95%
2-(Piperidin-1-yl)pyrimidine-5-carbaldehyde
Formel: C₁₀H₁₃N₃O
Molecular Weight: 191,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06589847
CAS-Nummer: 149806-11-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR