1-Isopropyl-1H-pyrrole-3-carbaldehyde 95%
Lieferant: Apollo Scientific
OR919032-1G
OR919032-5G
APOSOR919032-1GEA
219
EUR
APOSOR919032-1G
APOSOR919032-5G
1-Isopropyl-1H-pyrrole-3-carbaldehyde 95%
1-Isopropyl-1H-pyrrole-3-carbaldehyde
Formel:
C₈H₁₁NO Molecular Weight: 137.179 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11695054 CAS-Nummer: 30186-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...