Fmoc-(S)-3-Cyclopentylalanine 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-cyclopentylpropanoic acid

OR919154-1G OR919154-250MG OR919154-500MG
APOSOR919154-1GEA 654 EUR
APOSOR919154-1G APOSOR919154-250MG APOSOR919154-500MG
Fmoc-(S)-3-Cyclopentylalanine 97%
Fmoc-(S)-3-Cyclopentylalanine
Formel: C₂₃H₂₅NO₄
Molecular Weight: 379.449 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094886
CAS-Nummer: 371770-32-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR