3-(Thiophen-3-yl)propan-1-ol 95%

Lieferant: Apollo Scientific

Synonyme: 3-(3-Thienyl)-1-propanol

OR919164-1G OR919164-250MG
APOSOR919164-1GEA 258 EUR
APOSOR919164-1G APOSOR919164-250MG
3-(Thiophen-3-yl)propan-1-ol 95%
3-(Thiophen-3-yl)propan-1-ol
Formel: C₇H₁₀OS
Molecular Weight: 142,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09927266
CAS-Nummer: 20905-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR