3-Pyridinebutanol 95%
Lieferant: Apollo Scientific
Synonyme:
4-(Pyridin-3-yl)butan-1-ol
OR919224-1G
OR919224-5G
APOSOR919224-1GEA
186
EUR
APOSOR919224-1G
APOSOR919224-5G
3-Pyridinebutanol 95%
3-Pyridinebutanol
Formel:
C₉H₁₃NO Molecular Weight: 151,206 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18802565 CAS-Nummer: 60753-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...