3-Amino-N,N-dimethylbenzylamine 96%

Lieferant: Apollo Scientific

Synonyme: 3-((Dimethylamino)methyl)aniline

OR919226-1G
APOSOR919226-1GEA 119 EUR
APOSOR919226-1G
3-Amino-N,N-dimethylbenzylamine 96%
3-Amino-N,N-dimethylbenzylamine
Formel: C₉H₁₄N₂
Molecular Weight: 150,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036095
CAS-Nummer: 27958-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR