(2-Chloro-thiazol-5-ylmethyl)-cyclopropyl-amine 95%

Lieferant: Apollo Scientific

Synonyme: N-((2-Chlorothiazol-5-yl)methyl)cyclopropanamine

OR919268-1G
APOSOR919268-1GEA 266 EUR
APOSOR919268-1G
(2-Chloro-thiazol-5-ylmethyl)-cyclopropyl-amine 95%
(2-Chloro-thiazol-5-ylmethyl)-cyclopropyl-amine
Formel: C₇H₉ClN₂S
Molecular Weight: 188,678 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18837200
CAS-Nummer: 1289388-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR