2-Amino-1-boc-imidazole 97%
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 2-aminoimidazole-1-carboxylate
OR919651-1G
OR919651-250MG
OR919651-5G
APOSOR919651-1GEA
236
EUR
APOSOR919651-1G
APOSOR919651-250MG
APOSOR919651-5G
2-Amino-1-boc-imidazole 97%
2-Amino-1-boc-imidazole
Formel:
C₈H₁₃N₃O₂ Molecular Weight: 183.208 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20702858 CAS-Nummer: 929568-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...