2-(5-Ethynylthien-2-yl)pyridine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Ethynyl-5-(pyridin-2-yl)thiophene

OR9198-1G OR9198-250MG OR9198-50MG
APOSOR9198-1GEA 483 EUR
APOSOR9198-1G APOSOR9198-250MG APOSOR9198-50MG
2-(5-Ethynylthien-2-yl)pyridine 97%
2-(5-Ethynylthien-2-yl)pyridine
Formel: C₁₁H₇NS
Molecular Weight: 185,245 g/mol
Schmelzpunkt: 109…111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08271876
CAS-Nummer: 132464-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR