3-(Methoxymethoxy)aniline 95%
Lieferant: Apollo Scientific
OR919866-1G
OR919866-5G
APOSOR919866-1GEA
319
EUR
APOSOR919866-1G
APOSOR919866-5G
3-(Methoxymethoxy)aniline 95%
3-(Methoxymethoxy)aniline
Formel:
C₈H₁₁NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD21195211 CAS-Nummer: 96649-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...