7,8-Dihydro-5h-pyrano[4,3-c]pyridazin-3-ol 95%

Lieferant: Apollo Scientific

OR919889-1G OR919889-250MG
APOSOR919889-1GEA 647 EUR
APOSOR919889-1G APOSOR919889-250MG
7,8-Dihydro-5h-pyrano[4,3-c]pyridazin-3-ol 95%
7,8-Dihydro-5h-pyrano[4,3-c]pyridazin-3-ol
Formel: C₇H₈N₂O₂
Molecular Weight: 152,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26131634
CAS-Nummer: 1075260-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR