2-Methyl-l-proline methyl ester hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: (S)-Methyl 2-methylpyrrolidine-2-carboxylate hydrochloride

OR919993-1G OR919993-5G
APOSOR919993-1GEA 31.9 EUR
APOSOR919993-1G APOSOR919993-5G
2-Methyl-l-proline methyl ester hydrochloride 95%
2-Methyl-l-proline methyl ester hydrochloride
Formel: C₇H₁₃NO₂·ClH
Molecular Weight: 179,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22570171
CAS-Nummer: 220060-08-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR