5-(4-Ethynylphenyl)-1,3-oxazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(1,3-Oxazol-5-yl)phenylacetylene
OR9200-1G
OR9200-250MG
APOSOR9200-1GEA
452
EUR
APOSOR9200-1G
APOSOR9200-250MG
5-(4-Ethynylphenyl)-1,3-oxazole 97%
5-(4-Ethynylphenyl)-1,3-oxazole
Formel:
C₁₁H₇NO Molecular Weight: 169,179 g/mol Schmelzpunkt: 75,5…77.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08435847 CAS-Nummer: 501944-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...