Benzyl 2-hydroxy-2-methylpropionate 97%

Lieferant: Apollo Scientific

Synonyme: Benzyl 2-hydroxy-2-methylpropanoate

OR920076-1G OR920076-250MG OR920076-5G
APOSOR920076-1GEA 52.2 EUR
APOSOR920076-1G APOSOR920076-250MG APOSOR920076-5G
Benzyl 2-hydroxy-2-methylpropionate 97%
Benzyl 2-hydroxy-2-methylpropionate
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21641337
CAS-Nummer: 19444-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR