3,8-Diazabicyclo[3.2.1]octane dihydrochloride 95%

Lieferant: Apollo Scientific

OR920584-1G OR920584-250MG
APOSOR920584-1GEA 319 EUR
APOSOR920584-1G APOSOR920584-250MG
3,8-Diazabicyclo[3.2.1]octane dihydrochloride 95%
3,8-Diazabicyclo[3.2.1]octane dihydrochloride
Formel: C₆H₁₂N₂·2ClH
Molecular Weight: 185,095 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22689350
CAS-Nummer: 90673-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR