8-Chloro-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one 98%
Lieferant: Apollo Scientific
OR920734-1G
OR920734-250MG
APOSOR920734-1GEA
591
EUR
APOSOR920734-1G
APOSOR920734-250MG
8-Chloro-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one 98%
8-Chloro-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one
Formel:
C₆H₄ClN₃O Molecular Weight: 169,568 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21640309 CAS-Nummer: 1020042-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...