5-Methoxy-2,3-dihydro-1H-indene-1-carboxylic acid 95%

Lieferant: Apollo Scientific

OR920735-1G OR920735-250MG
APOSOR920735-1GEA 864 EUR
APOSOR920735-1G APOSOR920735-250MG
5-Methoxy-2,3-dihydro-1H-indene-1-carboxylic acid 95%
5-Methoxy-2,3-dihydro-1H-indene-1-carboxylic acid
Formel: C₁₁H₁₂O₃
Molecular Weight: 192,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07778334
CAS-Nummer: 116854-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR