((Cyclopent-3-en-1-yloxy)methyl)benzene 95%

Lieferant: Apollo Scientific

OR920740-1G OR920740-250MG
APOSOR920740-1GEA 696 EUR
APOSOR920740-1G APOSOR920740-250MG
((Cyclopent-3-en-1-yloxy)methyl)benzene 95%
((Cyclopent-3-en-1-yloxy)methyl)benzene
Formel: C₁₂H₁₄O
Molecular Weight: 174.239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20483839
CAS-Nummer: 37005-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR