Boc-L-glutamine methyl ester 97%
Lieferant: Apollo Scientific
Synonyme:
Methyl (2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoate
OR920855-1G
OR920855-5G
APOSOR920855-1GEA
151
EUR
APOSOR920855-1G
APOSOR920855-5G
Boc-L-glutamine methyl ester 97%
Boc-L-glutamine methyl ester
Formel:
C₁₁H₂₀N₂O₅ Lagertemperatur: Kühlschrank |
MDL:
MFCD25541921 CAS-Nummer: 4976-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...