sec-Butyl methanesulphonate 96%

Lieferant: Apollo Scientific

OR921213-1G OR921213-5G
APOSOR921213-1GEA 270 EUR
APOSOR921213-1G APOSOR921213-5G
sec-Butyl methanesulphonate 96%
sec-Butyl methanesulphonate
Formel: C₅H₁₂O₃S
Molecular Weight: 152,212 g/mol
Siedepunkt: 82 °C (2 mm Hg)
Dichte: 1,1053 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD01715284
CAS-Nummer: 16156-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR