1H-Pyrrolo[3,2-c]pyridine-3-carbonitrile 95%

Lieferant: Apollo Scientific

OR921315-1G OR921315-250MG
APOSOR921315-1GEA 185 EUR
APOSOR921315-1G APOSOR921315-250MG
1H-Pyrrolo[3,2-c]pyridine-3-carbonitrile 95%
1H-Pyrrolo[3,2-c]pyridine-3-carbonitrile
Formel: C₈H₅N₃
Molecular Weight: 143.145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18380871
CAS-Nummer: 1260385-57-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR