3-Chloro-7-nitro-1H-indazole 95%

Lieferant: Apollo Scientific

OR921456-1G OR921456-5G
APOSOR921456-1GEA 219 EUR
APOSOR921456-1G APOSOR921456-5G
3-Chloro-7-nitro-1H-indazole 95%
3-Chloro-7-nitro-1H-indazole
Formel: C₇H₄ClN₃O₂
Molecular Weight: 197,579 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11976299
CAS-Nummer: 74209-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR