(R)-N,N-Dimethyl-2-pyrrolidinecarboxamide hydrochloride 97%

Lieferant: Apollo Scientific

OR921459-1G OR921459-500MG
APOSOR921459-1GEA 522 EUR
APOSOR921459-1G APOSOR921459-500MG
(R)-N,N-Dimethyl-2-pyrrolidinecarboxamide hydrochloride 97%
(R)-N,N-Dimethyl-2-pyrrolidinecarboxamide hydrochloride
Formel: C₇H₁₄N₂O·ClH
Molecular Weight: 178,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28891479
CAS-Nummer: 566159-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR