4-Bromo-2-methylpyridin-3-amine 95%

Lieferant: Apollo Scientific

OR921486-100MG OR921486-1G OR921486-250MG
APOSOR921486-100MGEA 29 EUR
APOSOR921486-100MG APOSOR921486-1G APOSOR921486-250MG
4-Bromo-2-methylpyridin-3-amine 95%
4-Bromo-2-methylpyridin-3-amine
Formel: C₆H₇BrN₂
Molecular Weight: 187.037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16606103
CAS-Nummer: 126325-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR