3-Chlorocyclobutane-1-carboxylic acid
Lieferant: Apollo Scientific
Synonyme:
3-Chlorocyclobutanecarboxylic acid
OR921690-1G
OR921690-5G
APOSOR921690-1GEA
237
EUR
APOSOR921690-1G
APOSOR921690-5G
3-Chlorocyclobutane-1-carboxylic acid
3-Chlorocyclobutane-1-carboxylic acid
Formel:
C₅H₇ClO₂ Molecular Weight: 134.561 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07366750 CAS-Nummer: 35207-71-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...