Pyrrolidin-3-yl-ethanone hydrochloride 96%
Lieferant: Apollo Scientific
OR921754-1G
OR921754-250MG
APOSOR921754-1GEA
666
EUR
APOSOR921754-1G
APOSOR921754-250MG
Pyrrolidin-3-yl-ethanone hydrochloride 96%
Pyrrolidin-3-yl-ethanone hydrochloride
Formel:
C₆H₁₁NO·ClH Molecular Weight: 149.619 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18381078 CAS-Nummer: 876505-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...