5-Cyanonicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Cyanopyridine-3-carboxylic acid

OR921786-250MG OR921786-1G
APOSOR921786-250MGEA 122 EUR
APOSOR921786-250MG APOSOR921786-1G
5-Cyanonicotinic acid
5-Cyanonicotinic acid
Formel: C₇H₄N₂O₂
Molecular Weight: 148.119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07367904
CAS-Nummer: 887579-62-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR