(2R)-2-(Methoxymethyl)azetidine
Lieferant: Apollo Scientific
Synonyme:
(R)-2-(Methoxymethyl)azetidine
OR922129-1G
APOSOR922129-1GEA
687
EUR
APOSOR922129-1G
(2R)-2-(Methoxymethyl)azetidine
(2R)-2-(Methoxymethyl)azetidine
Formel:
C₅H₁₁NO Molecular Weight: 101,147 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11977227 CAS-Nummer: 935668-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...