3-Acetyl-4-chloro-7-azaindole 95%
Lieferant: Apollo Scientific
OR922295-1G
OR922295-250MG
APOSOR922295-1GEA
108
EUR
APOSOR922295-1G
APOSOR922295-250MG
3-Acetyl-4-chloro-7-azaindole 95%
3-Acetyl-4-chloro-7-azaindole
Formel:
C₉H₇ClN₂O Molecular Weight: 194,618 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12828645 CAS-Nummer: 1011711-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...