(S)-tert-Butyl 2-amino-3-phenylpropylcarbamate 97%

Lieferant: Apollo Scientific

Synonyme: (S)-tert-Butyl (2-amino-3-phenylpropyl)carbamate

OR922386-1G
APOSOR922386-1GEA 174 EUR
APOSOR922386-1G
(S)-tert-Butyl 2-amino-3-phenylpropylcarbamate 97%
(S)-tert-Butyl 2-amino-3-phenylpropylcarbamate
Formel: C₁₄H₂₂N₂O₂
Molecular Weight: 250,337 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11858348
CAS-Nummer: 167298-44-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR