N-BOC-erythro-L-β-Methylphenylalanine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S,3S)-Boc-β-methyl-L-Phe-OH

OR922464-100MG OR922464-25MG
APOSOR922464-100MGEA 91.5 EUR
APOSOR922464-100MG APOSOR922464-25MG
N-BOC-erythro-L-β-Methylphenylalanine 97%
N-BOC-erythro-L-β-Methylphenylalanine
Formel: C₁₅H₂₁NO₄
Molecular Weight: 279,332 g/mol
Schmelzpunkt: 101…105 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00800584
CAS-Nummer: 90731-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR