1-tert-Butylazetidin-3-amine

Lieferant: Apollo Scientific

OR922475-10G OR922475-1G OR922475-5G
APOSOR922475-10GEA 1040 EUR
APOSOR922475-10G APOSOR922475-1G APOSOR922475-5G
1-tert-Butylazetidin-3-amine
1-tert-Butylazetidin-3-amine
Formel: C₇H₁₆N₂
Molecular Weight: 128,215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10565786
CAS-Nummer: 18713-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR